Order Entry
Sweden
ContactUsLinkComponent
 
N,N-Dimethylformamide ≥99.5% (by GC)
 :  TCI
 
undefined
N,N-Dimethylformamide ≥99.5% (by GC)
 :  TCI

 

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %