Order Entry
Sweden
ContactUsLinkComponent
 
 
cis-2-Penten-1-ol ≥95.0% (by GC)
 :  TCI
 
undefined
 
cis-2-Penten-1-ol ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 95.0 %