(±)-2-Ethyl-1-hexanol ≥99.5% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:104-76-7
:
Formula:CH₃(CH₂)₃CH(C₂H₅)CH₂OH
MDL Number:MFCD00004746
:130.23 g/mol
Merck Index:14,03808
Boiling Pt:187°C
Melting Pt:–76 °C
Density:0,83 g/cm³ (20 °C)
Flash Pt:78°C
Storage Temperature:Ambient
EINECS:203-234-3
REACH:01-2119487289-20
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.5 %