Order Entry
Sweden
ContactUsLinkComponent
 
 
2-Bromopyrimidine ≥98.0%
 :  TCI
 
undefined
 
2-Bromopyrimidine ≥98.0%
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    52.0 to 58.0 deg-C