4-Fluoro-m-cresol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:452-70-0
:
Formula:C₇H₇FO
MDL Number:MFCD00042296
:126.13 g/mol
ADR:8,III
:GHS07
Specification Test Results
- Appearance:White or Colorles to Yellow to Orange powder to lump to clear liquid
- Purity (GC):min. 97.0 %