Order Entry
Sweden
ContactUsLinkComponent
 
 
 
 
3-Fluorophenyl isocyanate ≥97.0% (by GC)
  TCIAI0554-5G
 :  TCI
 :  
undefined
 
 
 
3-Fluorophenyl isocyanate ≥97.0% (by GC)
  TCIAI0554-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :137.11 g/mol
 :69°C
 :1,201 g/cm³ (20 °C)
 :42°C
 :Refrigerator
 :206-966-1
 :2478
 :3,III
 :GHS02
 :GHS06
 :GHS08
 :404-71-7
 :C₇H₄FNO
 :MFCD00002015