0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
1-Bromo-2,3,4-trifluorobenzene
1-Bromo-2,3,4-trifluorobenzene ≥98.0% (by GC)
1-Bromo-2,3,4-trifluorobenzene ≥98.0% (by GC)
:
TCI
1-Bromo-2,3,4-trifluorobenzene ≥98.0% (by GC)
:
TCI
:
176317-02-5
:
:
C₆H₂BrF₃
:
MFCD00013248
:
210.98 g/mol
:
60°C
:
62°C
:
GHS07
Appearance:
Colorless to Light yellow clear liquid
Purity (GC):
min. 98.0 %
Manual Fluorination Reagents, Fluorinated Building Blocks