Order Entry
Sweden
ContactUsLinkComponent
 
 
 
 
4,5-Dichloro-2-nitroaniline ≥98.0% (by GC)
 :  TCI
 
undefined
 
 
 
4,5-Dichloro-2-nitroaniline ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    177.0 to 181.0 deg-C