Order Entry
Sweden
ContactUsLinkComponent
 
 
(R)-(-)-Hydratropic acid ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
(R)-(-)-Hydratropic acid ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %
  • Optical purity (GC):
    min. 98.0 ee%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Freezing point:
    27.0 to 30.0 deg-C