
(R)-(-)-1-Aminopropan-2-ol ≥97.0% (by GC, titration analysis)
: TCI
:2799-16-8
:
:C₃H₉NO
:MFCD00064428
:75.11 g/mol
:160°C
:24…26 °C
:0,95 g/cm³ (20 °C)
:Kylskåp
:220-532-9
:3259
:8,II
:GHS05
- Appearance:White or Colorles to Yellow to Orange powder to lump to clear liquid
- Purity (GC):min. 97.0 %
- Optical purity (GC):min. 98.0 ee%
- Purity (Nonaqueous Titration):min. 97.0 %