Benzotriazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:95-14-7
:
Formula:C₆H₅N₃
MDL Number:MFCD00005699
:119.13 g/mol
Merck Index:14,01108
Boiling Pt:350 °C (1013 hPa)
Melting Pt:99 °C
Density:1,34 g/cm³ (20 °C)
Flash Pt: (closed cup)
Storage Temperature:Ambient
EINECS:202-394-1
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:96.0 to 100.0 deg-C
- Solubility in Methanol:almost transparency