tert-Butyl methyl ether ≥99.0% (by GC)
: TCI
:1634-04-4
:
:(CH₃)₃COCH₃
:MFCD00008812
:88.15 g/mol
:14,06032
:55°C
:–108.6 °C
:0,7405 g/cm³ (20 °C)
:–28°C
:Ambient
:216-653-1
:2398
:3,II
:GHS02
:GHS07
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %