Order Entry
Sweden
ContactUsLinkComponent
Sulphanilamide ≥99.0% (by titrimetric analysis) for biochemical research
Sulphanilamide ≥99.0% (by titrimetric analysis) for biochemical research
Catalog # TCIAS0833-5G
Supplier:  TCI
CAS Number:  
undefined
Sulphanilamide ≥99.0% (by titrimetric analysis) for biochemical research
Catalog # TCIAS0833-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (Diazo Method):
    min. 99.0 %
  • Nitrite ion detection test (Griess method):
    to pass test

Specification Test Results

About this item

 :
 :172.21 g/mol
Merck Index:14,08925
Boiling Pt:401 °C (1013 hPa)
Melting Pt:163…167 °C
Density:1,46 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-563-4
 :GHS07
CAS Number:63-74-1
Formula:H₂NC₆H₄SO₂NH₂
MDL Number:MFCD00007939