Order Entry
Sweden
ContactUsLinkComponent
 
Sulphanilamide ≥99.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
Sulphanilamide ≥99.0% (by HPLC, titration analysis)
 :  TCI
 

 

  • Appearance:
    White powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (Diazo Method):
    min. 99.0 %
  • Solubility in Sodium hydroxide solution:
    almost transparency