

(±)-2-Mercaptopropionic acid ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:79-42-5
:
Formula:C₃H₆O₂S
MDL Number:MFCD00004862
:106.15 g/mol
Merck Index:14,09340
Boiling Pt:115°C
Melting Pt:10…14 °C
Density:1,2 g/cm³ (20 °C)
Flash Pt:107°C
EINECS:201-206-5
UN:2936
ADR:6.1,II
:GHS05
:GHS06
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
- Purity (Iodometric Titration):min. 95.0 %