Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(1H-1,2,4-Triazol-1-yl)aniline
4-(1H-1,2,4-Triazol-1-yl)aniline
4-(1H-1,2,4-Triazol-1-yl)aniline
4-(1H-1,2,4-Triazol-1-yl)aniline
Leverantör:
Thermo Scientific
Certifikat
Expand image
4-(1H-1,2,4-Triazol-1-yl)aniline
Leverantör:
Thermo Scientific
Om denna produkt
CAS-nummer:
6523-49-5
:
Formel:
C₈H₈N₄
MDL-nummer:
MFCD00171470
:
160.1759 g/mol