
4-(4-Nitrophenylazo)resorcinol 90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:74-39-5
:
Formula:C₁₂H₉N₃O₄
MDL Number:MFCD00007310
:259.22 g/mol
Melting Pt:185 °C (dec.)
EINECS:200-808-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Orange-red to red Powder
- Infrared spectrum:Conforms
- HPLC:≥88 %
- Specific extinction E (1%/1cm):≥1400 (554 to 559 nm, 1N NaOH)