Order Entry
Sweden
ContactUsLinkComponent
Tetrachlorobisphenol A
Tetrachlorobisphenol A
Catalog # ACCUFRS022N
Supplier:  AccuStandard
CAS Number:  
Tetrachlorobisphenol A
Catalog # ACCUFRS022N
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Förpackningstyp:
    Glass bottle for solids
  • Förp.:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
EINECS:201-237-4
CAS Number:79-95-8
Formula:C₁₅H₁₂Cl₄O₂
MDL Number:MFCD00002178