Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Molybdenum carbide
Molybdenum carbide ≥99.5%
Molybdenum carbide ≥99.5%
Molybdenum carbide ≥99.5%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Molybdenum carbide ≥99.5%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
12069-89-5
:
Formula:
CMo₂
MDL Number:
MFCD00014218
:
203.89 g/mol
Melting Pt:
2687 °C
Storage Temperature:
Omgivande
EINECS:
235-115-7
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA012192
Videos
Thermo Scientific Acroseal 25 Years
Documentation