Order Entry
Sweden
ContactUsLinkComponent
Spectra/Por® Micro-dialysis Hollow Fibers
Spectra/Por® Micro-dialysis Hollow Fibers
Catalog # SPEC132294
Spectra/Por® Micro-dialysis Hollow Fibers
Catalog # SPEC132294
Specifications
  • Description:
    Micro-dialysis hollow fiber membranes, in vivo, cellulose, 152,4 mm
  • Pk:
    20
Specifications
About this item
The implantation of individual micro-dialysis hollow fibers is an ideal step in the recovery or introduction of micro-quantity biologicals or pharmaceutical materials in a localized tissue environment. Following sample injection into the lumen via syringe, the fiber ends may be sealed with epoxy for implantation.
Supplied in cut lengths of 152 mm (6").