



2,2,3-Trimethylbutane ≥96.0% (by GC)
: TCI
:464-06-2
:
:C₇H₁₆
:MFCD00039846
:100.2 g/mol
:81°C
:–24°C
:0,6901 g/cm³ (20 °C)
:–20°C
:207-346-3
:1206
:3,II
:GHS02
:GHS07
:GHS08
:GHS09
:Ambient
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %