Order Entry
Sweden
ContactUsLinkComponent
 
1-Triacontanol ≥85.0% (by GC)
 :  TCI
 
undefined
1-Triacontanol ≥85.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 85.0 %
  • Melting point:
    85.0 to 90.0 deg-C