Volumetric spoons, disposable, SteriWare®
These volumetric spoons allow small volumes to be measured accurately. It made of HDPE and are designed to be used straight from the bag.
- Moulded and packed in a cleanroom
- BSE/TSE free
- Full Batch Traceability
- Fully validated sterilisation procedure. SAL 10⁻⁶
- Available in a range of sizes from 1 to 20 ml
Suitable for food contact: FDA & EU 10/2011 conforming materials.
: Individually bagged.