Order Entry
Sweden
ContactUsLinkComponent
Disposable particle filtering respirators, FFP2, Aura™ 9400+
Disposable particle filtering respirators, FFP2, Aura™ 9400+
Katalog # 111-0718
Supplier:  3M
undefined
Disposable particle filtering respirators, FFP2, Aura™ 9400+
Katalog # 111-0718
Supplier:  3M

Specifications

  • Type:
    9422+
  • Pk:
    10
  • With valve:
    +
  • Class:
    FFP2D

Specifications

About this item

Three-panel design fits a wide range of face shapes and sizes to accommodate individual facial movements.

  • Staple-free: Helps to minimise contamination risks
  • Metal detectable: Embedded metal nose clip to help ensure production safety
  • Concealed adjustable nose clip: Kept securely in place within the layers of the respirator
  • Cool Flow™ valve effectively removes heat build-up, removes exhaled air and minimises the risk of fogging eyewear
  • Flat-fold; easy to store when not in use

Colour: Blue

Certifications: EN 149:2001+A1:2009
Caution: *One size fits all