Order Entry
Sweden
ContactUsLinkComponent
 
Fast red TR salt (1,5-naphthalenedisulphonate salt), Sigma-Aldrich®
 
undefined
Fast red TR salt (1,5-naphthalenedisulphonate salt), Sigma-Aldrich®

 

  • Appearance (Colour):
    White to Light Yellow
  • Appearance (Form):
    Powder
  • Proton NMR spectrum:
    Conforms to Structure
  • Solubility (Turbidity):
    Clear to Slightly Hazy
  • Solubility (Colour), c = 25 mg/ml, H₂O, Solvent Content (by GC):
    Faint Yellow to Light Yellow-Green
  • Suitability, Suitable as Coupling Agent with Naphthol AS:
    Suitable