

Rhodamine B isothiocyante mixture of isomers, powder, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:36877-69-7
:
Formula:C₂₉H₃₀ClN₃O₃S
MDL Number:MFCD00136007
:536.08 g/mol
ADR:9,III
:M-Clarity™ quality level = MQ200
EINECS:253-248-9
UN:3335
:GHS07
:GHS08
Storage Temperature:Freezer