Acriflavine hydrochloride
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69235-50-3
:8063-24-9
Formula:C₂₇H₂₈Cl₄N₆
MDL Number:MFCD00069039
Melting Pt:260 °C (dec.)
Storage Temperature:Ambient
UN:3077
ADR:9,III
:578.36 g/mol
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:Orange-red to red-brown Powder
- Infrared spectrum:Conforms
- Loss on drying:≤7 % (105°C, 1 h)