

Tetrachlorobis(tetrahydrofuran)titanium(IV) ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:31011-57-1
:
Formula:C₄H₈Cl₄OTi
MDL Number:MFCD00077884
:333.91 g/mol
Melting Pt:126…128 °C
Flash Pt:–17 °C (1 °F)
Storage Temperature:Ambient
UN:2925
ADR:4.1,II
:GHS02
:GHS05