Order Entry
Sweden
ContactUsLinkComponent
Monensin monosodium salt hydrate, Supelco®
Monensin monosodium salt hydrate, Supelco®
Catalog # 46468-100MG
Supplier:  Merck
CAS Number:  
undefined
Monensin monosodium salt hydrate, Supelco®
Catalog # 46468-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :692.85 g/mol (anhy)
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
EINECS:244-941-7
UN:3462
CAS Number:22373-78-0
Formula:C₃₆H₆₁NaO₁₁·H₂O
MDL Number:MFCD00077826