Order Entry
Sweden
ContactUsLinkComponent
Monensin monosodium salt hydrate
Monensin monosodium salt hydrate
Catalog # EHERC15291000
Supplier:  EHRENSTORFER
CAS Number:  
Monensin monosodium salt hydrate
Catalog # EHERC15291000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :692.85 g/mol (anhy)
EINECS:244-941-7
UN:3462
ADR:6.1,II
CAS Number:22373-78-0
Formula:C₃₆H₆₁NaO₁₁·H₂O
MDL Number:MFCD00077826