Order Entry
Sweden
ContactUsLinkComponent
8-Quinolineboronic acid
8-Quinolineboronic acid
Catalog # MOLEM10809364
Supplier:  Molekula
CAS Number:  
8-Quinolineboronic acid
Catalog # MOLEM10809364
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :172.98 g/mol
CAS Number:86-58-8
Formula:C₉H₈BNO₂
MDL Number:MFCD01114698