
Fast black K salt, Practical grade, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:64071-86-9
:
Formula:C₂₈H₂₄Cl₄N₁₀O₈Zn
MDL Number:MFCD00078471
Melting Pt:150 °C
:M-Clarity™ quality level = MQ200
EINECS:264-656-1
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (colour):Brown or red
- Appearance (form):Powder
- Solubility (colour):10 mg/ml water
- Solubility (method):10 mg/ml water
- Carbon content:9.9 - 18.3 %
- Nitrogen content:4.1 - 7.6 %
- Dye content:≥ 25 %