Creatine (N-amidinosarcosine) monohydrate
Catalog # MOLE52044294-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:149.15 g/mol (131.13 anhy)
EINECS:200-306-6
CAS Number:6020-87-7
Formula:C₄H₉N₃O₂·1H₂O
MDL Number:MFCD00071582