Order Entry
Sweden
ContactUsLinkComponent
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥97.5% (by HPLC), Sigma-Aldrich®
Health
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥97.5% (by HPLC), Sigma-Aldrich®
Certificates
undefined
Health
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥97.5% (by HPLC), Sigma-Aldrich®

Specification Test Results

  • Appearance (colour):
    Yellow to very dark yellow and orange to very dark orange
  • Appearance (form):
    Powder
  • Purity (hplc area %):
    ≥97.5%
  • Solubility (colour):
    Yellow to very dark yellow and green-yellow to very dark green-yellow
  • Solubility (turbidity):
    Clear
  • Solubility (method):
    10 mg/ml acetone
  • Infrared spectrum:
    Conforms to structure
  • Recommended retest period:
    45 months

Specification Test Results

Frequently Bought Together