
Brilliant green, crystals
Catalog # 720-0185
Supplier: RAL
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:482.65 g/mol
EINECS:211-190-1
:GHS07
Storage Temperature:Ambient
CAS Number:633-03-4
Formula:C₂₇H₃₄N₂O₄S
MDL Number:MFCD00011880