N-Acetylglycine(Ac-Gly-OH) 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:543-24-8
:
Formula:C₄H₇NO₃
MDL Number:MFCD00004275
:117.1 g/mol
Melting Pt:207…209 °C
Storage Temperature:Ambient
EINECS:208-839-6
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- Sulfated ash:≤0.1 %
- Titration with NaOH:≥98.5 % (On dry substance)
- Impurity:≤0.25 % Glycine
- Sulfate (SO4):≤0.2 %