Didecyldimethylammonium chloride 50% in 2-propanol 40% (v/v)/water 60% (v/v) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7173-51-5
:
Formula:C₂₂H₄₈ClN
MDL Number:MFCD00066262
:362.08 g/mol
ADR:8 (3),II
:M-Clarity™ quality level = MQ200
EINECS:230-525-2
UN:2920
:GHS02
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Boiling Point:88 °C (1013 hPa)
- Flash Point:29 °C
- Density:0.90 g/cm3 (20 °C)
- pH:6.5 - 8.0 (10 g/l, H2O, 20 °C)