Order Entry
Sweden
Orders LinkContactUsLinkComponent
AMPSO

AMPSO

Glass bottle for solids, 100 g
Catalog # MOLE17283132-100G
Marketsource
Supplier:   Molekula
AMPSO
Catalog # MOLE17283132-100G
Supplier:  Molekula
Marketsource
CAS Number:  68399-79-1
AMPSO

Glass bottle for solids, 100 g

CAS Number:  68399-79-1
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulphonic acid
  • CAS Number:68399-79-1
  • Formula:C₇H₁₇NO₅S
  • MDL Number:MFCD00041777
  • Molecular Weight:227.28 g/mol
  • Storage Temperature:Ambient
  • EINECS:269-991-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.