DIPSO
Glass bottle for solids, 100 g
Catalog # MOLE34218155-100G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
- CAS Number:68399-80-4
- Formula:C₇H₁₇NO₆S
- MDL Number:MFCD00038353
- Molecular Weight:261.29 g/mol
- Storage Temperature:Omgivande
- EINECS:269-992-2
Specifications
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:100 g



