Safety goggles, Fahrenheit™
: 3M
3M™ Fahrenheit™ safety goggles feature a soft crystal frame and a wide support area around the face. They are designed with a specific channel on the side of the frame to accommodate the temple arms of prescription glasses.
- Economical large profile safety goggle range, with enough space to fit over most prescription glassess
- Polycarbonate lenses for impact resistance
- 3M 1 BT lens marking
- Indirect ventilation to protect against liquid droplets and large dust particles
- Cylindrical curved lens design for optimal horizontal field of vision
Get comfortable protection over the top of prescription glasses with 3M™ Fahrenheit™ safety goggles. These goggles feature a soft crystal frame with a wide support area around the face designed to enhance comfort. A specific channel on the side of the frame helps accommodate the temple arms of prescription frames, making them ideal for use with prescription glasses.
Weight: 68 g
: EN 166.
: Supplied with microfibre pouch.