Accessories for safety helmets
Catalog # DALL1002337
Supplier: HONEYWELL SAFETY
Specifications
- Description:Metal mesh for helmet attachment
- Pk:1
Specifications
About this item
Face protection visor or mesh as a helmet attachment.
- Electrician's faceshield: with clear PC visor (240 mm); protects against flying sparks and electrical arcs in the event of short circuits and chemical hazards; suitable for helmet with 30 mm slots
- SAM9 metal wire visor: visor made out of metal mesh for the Supervizor to protect against molten metal and radiated heat