Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Proteinase K
Proteinase K
Proteinase K
Supplier:
Ambion
Certificates
Expand image
Proteinase K
Supplier:
Ambion
About this item
Specifications
CAS No.:
39450-01-6
Storage conditions:
Refrigerator
Form:
Aqueous solution
Enzyme name:
Proteinase K
Enzyme activity:
600 mAnsonU/ml
Concentration:
20 mg/ml
Specifications