Order Entry
Sweden
ContactUsLinkComponent
Bovine Ribonuclease A (from Pancreas)
Bovine Ribonuclease A (from Pancreas)
Catalog # ROTH7164.2
Supplier:  Roth Carl
CAS Number:  
Bovine Ribonuclease A (from Pancreas)
Catalog # ROTH7164.2
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • CAS No.:
    9001-99-4
  • Source:
    Pancreas
  • Species:
    Bovine
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00132184
  • Enzyme name:
    Ribonuclease A
  • Enzyme activity:
    70 U/mg (Kunitz)
  • Enzyme synonyms:
    RNase A

Specifications

About this item