Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Lysozyme (Muramidase)
Lysozyme (Muramidase)
Lysozyme (Muramidase)
Catalog # A4972.0010
Supplier:
PanReac AppliChem
CAS Number:
Certificates
Expand image
Lysozyme (Muramidase)
Catalog # A4972.0010
Supplier:
PanReac AppliChem
CAS Number:
Certificates
Specifications
Pack type:
Glass bottle for solids
Pk:
10 g
CAS No.:
9001-63-2
Enzyme name:
Lysozyme (Muramidase)
Specifications
About this item
Product Details
Documentation
Product Details
Documentation