

tert-Butyl isocyanide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7188-38-7
:
Formula:C₅H₉N
MDL Number:MFCD00000002
:83.13 g/mol
Boiling Pt:91 °C
Density:0.73 g/cm³
Flash Pt:–2 °C
Storage Temperature:Refrigerator
EINECS:230-553-5
UN:1992
ADR:3,II
:GHS02
:GHS06
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3740 to 1.3790 (20°C, 589 nm)
- GC:≥96.0 %