Vitamin K1 (phytomenadione, phylloquinone), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:84-80-0
:81818-54-4
Formula:C₃₁H₄₆O₂
MDL Number:MFCD00214063
:450.6958 g/mol
EINECS:201-564-2