Pesticide reference standard, Flurprimidol 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL13851000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:56425-91-3
- Description:Flurprimidol 10 µg/ml in acetonitrile
- Formula:C₁₅H₁₅F₃N₂O₂
- Description:Flurprimidol