Pesticide reference standard, γ-Cyhalothrin 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL11859500CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:76703-62-3
- Description:γ-Cyhalothrin 10 µg/ml in cyclohexane
- Formula:C₂₃H₁₉ClF₃NO₃
- Description:γ-Cyhalothrin