Order Entry
Sweden
Orders LinkContactUsLinkComponent
2-(Ethylthio)nicotinamide

2-(Ethylthio)nicotinamide

Marketsource
Supplier:   Thermo Scientific
2-(Ethylthio)nicotinamide
Supplier:  Thermo Scientific
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
  • CAS Number:175135-27-0
  • Formula:C₈H₁₀N₂OS
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    MAYBBTB06390DA
    Marketsource
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    MAYBBTB06390EA
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.