Pesticide reference standard, 3,4,5-Trichlorophenol 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL17774800IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:609-19-8
- Description:3,4,5-Trichlorophenol 10 µg/ml in isooctane
- Formula:C₆H₃Cl₃O
- Description:3,4,5-Trichlorophenol